Learn More
Medchemexpress LLC 4-chlorophenylguanidine hydrochloride | 14279-91-5 | 99.1% | 206.08 g·mol⁻1 | C7H9Cl2N3 | 1 ML
SDP
Catalog No. 5000213331
Encompass_Preferred
4-Chlorophenylguanidine hydrochloride is a research-grade small molecule provided as solids and as a ready-to-use 10 mM solution in DMSO (1 mL). It is used in pharmacology and in vitro studies as an ASIC3 modulator and urokinase-related inhibitor. Chemical identifiers: CAS 14279-91-5; formula C7H9Cl2N3; molecular weight 206.08 g·mol⁻1.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Available as solid for custom concentration preparation.
  • High purity suitable for research applications.
  • Known chemical identifiers provided for traceability and reporting.
Catalog No. 50-002-13331 Supplier Medchemexpress LLC Supplier No. HY10093910MM 1ML
Add to Cart
Add to Cart
4-Chlorophenylguanidine hydrochloride is a research-grade small molecule provided as solids and as a ready-to-use 10 mM solution in DMSO (1 mL). It is used in pharmacology and in vitro studies as an ASIC3 modulator and urokinase-related inhibitor. Chemical identifiers: CAS 14279-91-5; formula C7H9Cl2N3; molecular weight 206.08 g·mol⁻1.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Available as solid for custom concentration preparation.
  • High purity suitable for research applications.
  • Known chemical identifiers provided for traceability and reporting.