Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylhydrazine Sulfate 95.0+%, TCI America™
SDP

Catalog No. C025825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025825G 25 g
C0258500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025825G Supplier TCI America Supplier No. C025825G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14581-21-6
Molecular Formula C6H7ClN2
MDL Number MFCD00043829

Specifications

Quantity 25 g
Formula Weight 383.24
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenylhydrazine Sulfate
EINECSNumber (3)-0471
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.