Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Chlorophenylsulfonyl)urea 97.0+%, TCI America™
SDP

Catalog No. C22665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22665G Supplier TCI America Supplier No. C22665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22663-37-2
Molecular Formula C7H7ClN2O3S
Molecular Weight (g/mol) 234.654
MDL Number MFCD00518656
InChI Key AZEPYUPSYWCRBG-UHFFFAOYSA-N
Synonym 4-chlorobenzenesulfonylurea, 4-chlorophenylsulfonyl urea, 1-4-chlorobenzenesulfonyl urea, 4-chlorobenzenesulfonyl urea, n-carbamoyl-4-chlorobenzenesulfonamide, p-chlorobenzenesulfonyl urea, p-chlorobenzene sulfonyl urea, n-aminocarbonyl-4-chlorobenzenesulphonamide, benzenesulfonamide, n-aminocarbonyl-4-chloro, amino-n-4-chlorophenyl sulfonyl amide
PubChem CID 89785
IUPAC Name (4-chlorophenyl)sulfonylurea
SMILES C1=CC(=CC=C1S(=O)(=O)NC(=O)N)Cl

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 234.65
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Chlorophenylsulfonyl)urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.