Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloropiperidine 97.0+%, TCI America™
SDP

Catalog No. C218225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C218225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C218225G Supplier TCI America Supplier No. C218225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5382-18-3
Molecular Formula C5H10ClN
Molecular Weight (g/mol) 119.592
InChI Key GBPBXBUHZSOKTH-UHFFFAOYSA-N
PubChem CID 420932
IUPAC Name 4-chloropiperidine
SMILES C1CNCCC1Cl

Specifications

Color Yellow
Boiling Point 60°C
Quantity 25 g
UN Number 2920
Formula Weight 119.59
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chloropiperidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.