Learn More
Accela Chembio Inc 4-chloroquinazoline | 25g | 5190-68-1 | MFCD00228682 | 97+% | Shelf Life: 1260 Days | Moisture Sensitive/air Sensitive/+4
Supplier:  Accela Chembio Inc SY00217325G
Encompass_Preferred
4-chloroquinazoline | 25g | 5190-68-1 | MFCD00228682 | 97+% | Shelf Life: 1260 Days | Moisture Sensitive/air Sensitive/+4
Catalog No. 50-353-338