Learn More
CELLTREAT Scientific Products 4 Chamber Cell Culture Slide, Sterile
SDP
Catalog No. 501149053
Encompass_Preferred
CELLTREAT Scientific Products 4 Chamber Cell Culture Slide. Grow cells directly on the slide without any need for future transfer; ideal for growing and viewing cells under a microscope on standard 25mm x 75mm slide. Integrated gasket prevents cross-contamination between chambers. Inert hydrophobic green border printed on slide to define cell culture area. Chamber holder features easy flip locks for assembly/disassembly; no separation tool required. Packing trays can be used as incubation racks in CO2 incubators. Uniform hydrophilic surface ideal for consistent cell attachment. Slides are labeled with numeric markings. Adhesive free. Made from glass. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 6/tray with 2 trays/box, 12/case.
Catalog No. 50-114-9053 Supplier CELLTREAT Scientific Products Supplier No. 229164
CELLTREAT Scientific Products 4 Chamber Cell Culture Slide. Grow cells directly on the slide without any need for future transfer; ideal for growing and viewing cells under a microscope on standard 25mm x 75mm slide. Integrated gasket prevents cross-contamination between chambers. Inert hydrophobic green border printed on slide to define cell culture area. Chamber holder features easy flip locks for assembly/disassembly; no separation tool required. Packing trays can be used as incubation racks in CO2 incubators. Uniform hydrophilic surface ideal for consistent cell attachment. Slides are labeled with numeric markings. Adhesive free. Made from glass. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 6/tray with 2 trays/box, 12/case.