Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-4-Cholesten-3-one, TCI America™
SDP

Catalog No. C13665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13665G Supplier TCI America Supplier No. C13665G

Chemical Identifiers

CAS 601-57-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
MDL Number MFCD00003663
InChI Key NYOXRYYXRWJDKP-GYKMGIIDSA-N
Synonym 4-cholesten-3-one, cholest-4-en-3-one, cholestenone, 3-oxocholest-4-ene, delta sup 4-cholestenone, +-4-cholesten-3-one, unii-7t94nhd99c, 3-oxo-4-cholestene, 3-keto-4-cholestene, cholestenone delta 4
PubChem CID 91477
ChEBI CHEBI:16175
IUPAC Name (8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=O)CCC34C)C

Specifications

Melting Point 81°C
Color Yellow
Quantity 5 g
Formula Weight 384.65
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-4-Cholesten-3-one
RTECSNumber FZ7700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.