Learn More
TARGETMOL CHEMICALS INC 4-CMTB 10MG
SDP
Catalog No. 502579058
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-CMTB is a selective agonist of FFA2 and GPR43.It also is a positive allosteric modulator (pEC50=6.38). purity: 99%
Catalog No. 50-257-9058 Supplier TARGETMOL CHEMICALS INC Supplier No. T864210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-CMTB is a selective agonist of FFA2 and GPR43.It also is a positive allosteric modulator (pEC50=6.38). purity: 99%