Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 95.0+%, TCI America™
SDP

Catalog No. C17821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17821G Supplier TCI America Supplier No. C17821G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38078-71-6
Molecular Formula C10H18N2O
Molecular Weight (g/mol) 182.27
MDL Number MFCD00191964
InChI Key KVTOSAMSZMXLKB-UHFFFAOYSA-N
Synonym 4-Cyano-TEMPO
PubChem CID 16212508
IUPAC Name 1-hydroxy-2,2,6,6-tetramethylpiperidine-4-carbonitrile
SMILES CC1(C)CC(CC(C)(C)N1O)C#N

Specifications

Melting Point 147°C
Quantity 1 g
Formula Weight 181.26
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.