Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-3-fluorobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. C27991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27991G Supplier TCI America Supplier No. C27991G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 176508-81-9
Melting Point 164°C
Color White
Molecular Formula C8H4FNO2
Quantity 1 g
MDL Number MFCD03094324
InChI Key ZWKNDLMYSLLMRF-UHFFFAOYSA-N
SMILES OC(=O)C1=CC(F)=C(C=C1)C#N
IUPAC Name 4-cyano-3-fluorobenzoic acid
Molecular Weight (g/mol) 165.12
PubChem CID 2778468
Formula Weight 165.12
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-3-fluorobenzoic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.