Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-3-fluorophenylboronic acid, 97%

Catalog No. AC448710050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448710050 5 g
AC448710010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448710050 Supplier Thermo Scientific Chemicals Supplier No. 448710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 843663-18-3 | C7H5BFNO2 | 164.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 843663-18-3
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03411549
InChI Key DECWLXUOZUMPBF-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug
PubChem CID 2783130
IUPAC Name (4-cyano-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(C=C1)C#N

Specifications

Melting Point 362°C to 366°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.93
Percent Purity 97%
Chemical Name or Material 4-Cyano-3-fluorophenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.