Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C24565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2456-5G 5 g
C24561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2456-5G Supplier TCI America Supplier No. C24565G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 843663-18-3
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03411549
InChI Key DECWLXUOZUMPBF-UHFFFAOYSA-N
PubChem CID 2783130
IUPAC Name (4-cyano-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(C=C1)C#N

Specifications

Color White-Yellow
Quantity 5 g
Synonym 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid
Formula Weight 164.93
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.