Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4'-dodecylbiphenyl 97.0+%, TCI America™
SDP

Catalog No. C32391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32391G 1 g
C32395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32391G Supplier TCI America Supplier No. C32391G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57125-49-2
Molecular Formula C25H33N
Molecular Weight (g/mol) 347.546
MDL Number MFCD08062573
InChI Key YTXPODSLSFRTBJ-UHFFFAOYSA-N
PubChem CID 92673
IUPAC Name 4-(4-dodecylphenyl)benzonitrile
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 48°C
Color White
Quantity 1 g
Synonym 4′C-Dodecyl-4-biphenylcarbonitrile, 12CB
Formula Weight 347.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-dodecylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.