Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Cyano-4-nonyloxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. C32721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32721G Supplier TCI America Supplier No. C32721G
 Restricted Item
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58932-13-1
Molecular Formula C22H27NO
Molecular Weight (g/mol) 321.464
MDL Number MFCD00388658
InChI Key NJQLLUIPSJLJHY-UHFFFAOYSA-N
Synonym 4′C-Nonyloxy-4-biphenylcarbonitrile, 4′C-(Nonyloxy)-[1,1′C-biphenyl]-4-carbonitrile, 9OCB
PubChem CID 100887
IUPAC Name 4-(4-nonoxyphenyl)benzonitrile
SMILES CCCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Color White
Quantity 1 g
Formula Weight 321.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Cyano-4-nonyloxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.