Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Cyano-4-phenylpiperidine hydrochloride

Catalog No. C9440810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C9440810G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C9440810G Supplier Sigma Aldrich Supplier No. C9440810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Cyano-4-phenylpiperidine hydrochloride

Specifications

CAS 51304-58-6
Melting Point 208°C to 210°C (lit.)
Molecular Formula C12H14N2 -+ HCl
Linear Formula C12H14N2 -+ HCl
MDL Number MFCD00012775
Quantity 250 mg
Synonym 4-Phenyl-4-piperidinecarbonitrile hydrochloride
Molecular Weight (g/mol) 222.71
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.