Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4'-propoxy-1,1'-biphenyl 98.0+%, TCI America™
SDP

Catalog No. C29115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29115G 5 g
C29111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29115G Supplier TCI America Supplier No. C29115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52709-86-1
Molecular Formula C16H15NO
Molecular Weight (g/mol) 237.302
MDL Number MFCD00183206
InChI Key RZCQJZFWJZHMPK-UHFFFAOYSA-N
Synonym 4'-propoxy-1,1'-biphenyl-4-carbonitrile, 3ocb, 1,1'-biphenyl-4-carbonitrile, 4'-propoxy, 4-propoxy-4'-cyanobiphenyl, 4-cyano-4'-propoxybiphenyl, 4-propoxycyanodiphenyl, 4-propoxy-1,1'-biphenyl-4'-carbonitrile, 4-4-propoxyphenyl benzonitrile, 4'-propoxybiphenyl-4-carbonitrile, 4'-propoxy-4-cyanobiphenyl
PubChem CID 104291
IUPAC Name 4-(4-propoxyphenyl)benzonitrile
SMILES CCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 73°C
Color White
Quantity 5 g
Formula Weight 237.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-propoxy-1,1′-biphenyl
RTECSNumber DV1764600
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.