Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-D-phenylalanine, 98%, Thermo Scientific™

Catalog No. AAH6368703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6368703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6368703 Supplier Thermo Scientific Chemicals Supplier No. H6368703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Cyano-D-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 263396-44-7
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD00270364
InChI Key KWIPUXXIFQQMKN-SECBINFHSA-N
Synonym r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cyanophenylalanine, h-d-phe 4-cn-oh, 4-cyano-d-phenylalanine, 2r-2-amino-3-4-cyanophenyl propanoic acid, d-4-cn-phe-oh, d-4-cyanophe, h-4-cyano-phe-oh, ambotzhaa7570, pubchem13160
PubChem CID 1501864
IUPAC Name (2R)-2-amino-3-(4-cyanophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)C#N

Specifications

Color White
UN Number UN3439
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 190.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Cyano-D-phenylalanine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.