Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanobenzoyl chloride, 98%

Catalog No. p-7027520
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2433306 5 g
AAB2433314 25 g
AAB2433322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2433306 Supplier Thermo Scientific Chemicals Supplier No. B2433306
Only null left
Add to Cart

CAS: 6068-72-0 | C8H4ClNO | 165.58 g/mol

It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.

Solubility
Reacts with water.

Notes
Store under dry inert gas. Keep away from moisture, oxidizing agents, acids, bases, active metals, amines and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6068-72-0
Molecular Formula C8H4ClNO
Molecular Weight (g/mol) 165.58
MDL Number MFCD00001822
InChI Key USEDMAWWQDFMFY-UHFFFAOYSA-N
Synonym 4-cyanobenzoylchloride, 4-cyano-benzoyl chloride, p-cyanobenzoyl chloride, unii-2577fde7a8, benzoyl chloride, 4-cyano, 4cyanobenzoyl chloride, 4-cyano-benzoylchloride, 4-cyanobenzoyl-chloride, 4-cyanpbenzoyl chloride, 4-cyano benzoylchloride
PubChem CID 80172
IUPAC Name 4-cyanobenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 68°C to 70°C
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 386729
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 165.58
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzoyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 228-005-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.