Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyanophenylhydrazine hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
2863-98-1
C7H8ClN3
169.612
MFCD00673994
UXDLLFIRCVPPQP-UHFFFAOYSA-N
4-cyanophenylhydrazine hydrochloride,4-hydrazinylbenzonitrile hydrochloride,4-hydrazinobenzonitrile hydrochloride,4-cyanophenylhydrazine hcl,benzonitrile, 4-hydrazino-, monohydrochloride,4-hydrazino-benzonitrile; hydrochloride,pubchem13684,4-cyanophenylhydrazine-hcl,acmc-1co7p,ksc205o1j
16212962
4-hydrazinylbenzonitrile;hydrochloride
C1=CC(=CC=C1C#N)NN.Cl
2863-98-1
C7H8ClN3
169.612
MFCD00673994
UXDLLFIRCVPPQP-UHFFFAOYSA-N
4-cyanophenylhydrazine hydrochloride,4-hydrazinylbenzonitrile hydrochloride,4-hydrazinobenzonitrile hydrochloride,4-cyanophenylhydrazine hcl,benzonitrile, 4-hydrazino-, monohydrochloride,4-hydrazino-benzonitrile; hydrochloride,pubchem13684,4-cyanophenylhydrazine-hcl,acmc-1co7p,ksc205o1j
16212962
4-hydrazinylbenzonitrile;hydrochloride
C1=CC(=CC=C1C#N)NN.Cl
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Melting Point ~243°C (decomposition)
Quantity 100g
Beilstein 3565486
Formula Weight 169.61
Percent Purity 97%
Chemical Name or Material 4-Cyanophenylhydrazine hydrochloride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger