Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyclohexylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. C04975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04975G 5 g
C049725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C04975G Supplier TCI America Supplier No. C04975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2433-14-9
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.31
MDL Number MFCD00060071,MFCD13185038
InChI Key AFKMHDZOVNDWLO-UHFFFAOYSA-N
PubChem CID 75518
IUPAC Name [1,1'-bi(cyclohexane)]-4-ol
SMILES OC1CCC(CC1)C1CCCCC1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 182.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexylcyclohexanol (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.