Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-cyclopropyl-7-fluoro-2,3-dihydro-1λ6,2,4-benzothiadiazine 1,1-dioxide | 1204572-55-3 | MFCD31579820 | 99.7% | 242.27 g/mol | C10H11FN2O2S | 5 MG
SDP

Catalog No. 5000262726
Encompass_Preferred

BPAM344 is a small-molecule positive allosteric modulator selective for kainate receptor subunits GluK1b, GluK2a, and GluK3a. Supplied as a milligram-scale research reagent, it is characterized by documented physicochemical data and high purity for use in in vitro pharmacology and receptor modulation studies.

  • High purity (99.7%).
  • Molecular formula C10H11FN2O2S.
  • Molecular weight 242.27 g/mol.
  • Soluble in DMSO at ≥5 mg/mL.
  • Offered in multiple milligram pack sizes for laboratory research.
  • Intended for in vitro studies of kainate receptor modulation.

Catalog No. 50-002-62726 Supplier Medchemexpress LLC Supplier No. HY1290865MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BPAM344 is a small-molecule positive allosteric modulator selective for kainate receptor subunits GluK1b, GluK2a, and GluK3a. Supplied as a milligram-scale research reagent, it is characterized by documented physicochemical data and high purity for use in in vitro pharmacology and receptor modulation studies.

  • High purity (99.7%).
  • Molecular formula C10H11FN2O2S.
  • Molecular weight 242.27 g/mol.
  • Soluble in DMSO at ≥5 mg/mL.
  • Offered in multiple milligram pack sizes for laboratory research.
  • Intended for in vitro studies of kainate receptor modulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.