Learn More
Cayman Chemical 4-DafachronIc AcId 10mg
SDP
Catalog No. 502955488
Encompass_Preferred
A racemic mixture of (25R)-Δ4-dafachronic acid and (25S)-Δ4-dafachronic acid; an agonist of DAF-12; a metabolite of cholesterol in C. elegans; induces transactivation of DAF-12 in NIH3T3 cells expressing the C. elegans or D. immitis receptor (EC50s = 81 and 0.65 nM, respectively); induces molting of infective D. immitis (heartworm) third-stage larvae into L4 larvae; induces the escape trajectory in embryos of the annual killifish A. limnaeus at 25°C at 75 µM
Catalog No. 50-295-5488 Supplier Cayman Chemical Supplier No. 1410010 MG
A racemic mixture of (25R)-Δ4-dafachronic acid and (25S)-Δ4-dafachronic acid; an agonist of DAF-12; a metabolite of cholesterol in C. elegans; induces transactivation of DAF-12 in NIH3T3 cells expressing the C. elegans or D. immitis receptor (EC50s = 81 and 0.65 nM, respectively); induces molting of infective D. immitis (heartworm) third-stage larvae into L4 larvae; induces the escape trajectory in embryos of the annual killifish A. limnaeus at 25°C at 75 µM