Learn More
Cayman Chemical Δ4-Dafachronic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502991534
Encompass_Preferred
An internal standard for the quantification of Δ4-dafachronic acid by GC- or LC-MS
Catalog No. 50-299-1534 Supplier Cayman Chemical Supplier No. 310091 MG
An internal standard for the quantification of Δ4-dafachronic acid by GC- or LC-MS