Learn More
Enzo Life Sciences 4-DAMP (50mg). CAS: 1952-15-4
SDP
Supplier:  Enzo Life Sciences BMLC1040050
Encompass_Preferred
M3 muscarinic receptor antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO and warm water. Long Term Storage: Ambient.
Catalog No. 50-201-0341