Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-decanol | 2051-31-2 | MFCD00039627 | 98.2% | 158.28 g/mol | C10H22O | 1 G
SDP

Catalog No. 5000282435
Encompass_Preferred

4-Decanol is a primary alcohol used as a research reagent with reported antimutagenic activity. It is typically employed in biochemical studies investigating mutagenesis inhibition and related cellular responses. Follow SDS and laboratory safety procedures when handling this compound.

  • Antimutagenic compound reported to inhibit aflatoxin B1 and MNNG mutagenicity.
  • Chemical formula C10H22O; molecular weight 158.28 g/mol.
  • High purity (98.2%).
  • CAS number 2051-31-2.
  • Available in small research quantities (1 g, 5 g, 10 g, 50 g).
  • Store under recommended conditions to maintain stability.

Catalog No. 50-002-82435 Supplier Medchemexpress LLC Supplier No. HYW2755531G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Decanol is a primary alcohol used as a research reagent with reported antimutagenic activity. It is typically employed in biochemical studies investigating mutagenesis inhibition and related cellular responses. Follow SDS and laboratory safety procedures when handling this compound.

  • Antimutagenic compound reported to inhibit aflatoxin B1 and MNNG mutagenicity.
  • Chemical formula C10H22O; molecular weight 158.28 g/mol.
  • High purity (98.2%).
  • CAS number 2051-31-2.
  • Available in small research quantities (1 g, 5 g, 10 g, 50 g).
  • Store under recommended conditions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.