Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-decanol | 2051-31-2 | MFCD00039627 | 98.2% | 158.28 g/mol | C10H22O | 1 ML
SDP

Catalog No. 5000282017
Encompass_Preferred

4-Decanol is a small organic alcohol used in research as an antimutagenic compound. It is supplied both as a ready-to-use 10 mM solution in DMSO and as solid material for analytical and experimental applications.

  • Antimutagenic small molecule used in in vitro assays.
  • Available as a 10 mM solution in DMSO and as solids (1 G, 5 G, 10 G, larger quantities by quote).
  • Purity 98.2%.
  • Molecular formula C10H22O and molecular weight 158.28 g/mol.
  • Highly soluble in DMSO (≥100 MG/ML); use freshly opened DMSO due to hygroscopicity.
  • Storage recommendations: pure form -20°C for up to 3 years; in solvent store at -80°C for long-term.

Catalog No. 50-002-82017 Supplier Medchemexpress LLC Supplier No. HYW27555310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Decanol is a small organic alcohol used in research as an antimutagenic compound. It is supplied both as a ready-to-use 10 mM solution in DMSO and as solid material for analytical and experimental applications.

  • Antimutagenic small molecule used in in vitro assays.
  • Available as a 10 mM solution in DMSO and as solids (1 G, 5 G, 10 G, larger quantities by quote).
  • Purity 98.2%.
  • Molecular formula C10H22O and molecular weight 158.28 g/mol.
  • Highly soluble in DMSO (≥100 MG/ML); use freshly opened DMSO due to hygroscopicity.
  • Storage recommendations: pure form -20°C for up to 3 years; in solvent store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.