Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-decanol | 2051-31-2 | MFCD00039627 | 98.2% | 158.28 g/mol | C10H22O | 10 G
SDP

Catalog No. 5000281305
Encompass_Preferred

4-decanol is a medium-chain fatty alcohol used as a research reagent with reported antimutagenic activity. It is supplied as a neat liquid and as a DMSO solution for biochemical assays; intended for research use only and not for human or veterinary use.

  • Antimutagenic fatty alcohol with reported inhibition of aflatoxin B1 and MNNG mutagenicity.
  • Purity 98.2%.
  • CAS 2051-31-2; molecular weight 158.28 g/mol; formula C10H22O.
  • Appearance: liquid, colorless to light yellow.
  • Available forms and sizes: 10 mM (1 mL in DMSO), 1 G, 5 G, 10 G, 50 G (bulk quote available).
  • Storage guidance: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-81305 Supplier Medchemexpress LLC Supplier No. HYW27555310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-decanol is a medium-chain fatty alcohol used as a research reagent with reported antimutagenic activity. It is supplied as a neat liquid and as a DMSO solution for biochemical assays; intended for research use only and not for human or veterinary use.

  • Antimutagenic fatty alcohol with reported inhibition of aflatoxin B1 and MNNG mutagenicity.
  • Purity 98.2%.
  • CAS 2051-31-2; molecular weight 158.28 g/mol; formula C10H22O.
  • Appearance: liquid, colorless to light yellow.
  • Available forms and sizes: 10 mM (1 mL in DMSO), 1 G, 5 G, 10 G, 50 G (bulk quote available).
  • Storage guidance: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.