Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Decenoic Acid 98.0+%, TCI America™
SDP

Catalog No. D44495ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44495ML 5 mL
D444925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44495ML Supplier TCI America Supplier No. D44495ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26303-90-2
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD03092963
InChI Key XKZKQTCECFWKBN-VOTSOKGWSA-N
PubChem CID 5282726
IUPAC Name (4E)-dec-4-enoic acid
SMILES CCCCC\C=C\CCC(O)=O

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Decenoic Acid
EINECSNumber (2)-0609
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.