Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Decyloxybenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. D112425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D112425G 25 g
D11245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D112425G Supplier TCI America Supplier No. D112425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24083-16-7
Molecular Formula C17H26O2
Molecular Weight (g/mol) 262.39
MDL Number MFCD00021072
InChI Key WOSYBKJRUQJISL-UHFFFAOYSA-N
PubChem CID 141068
IUPAC Name 4-(decyloxy)benzaldehyde
SMILES CCCCCCCCCCOC1=CC=C(C=O)C=C1

Specifications

Boiling Point 176°C
Quantity 25 g
Synonym 4-decyloxy benzaldehyde, 4-decyloxybenzaldehyde, p-decyloxybenzaldehyde, 4-n-decyloxybenzaldehyde, benzaldehyde,4-decyloxy, benzaldehyde, 4-decyloxy, 4-decylox-ybenzaldehyde, p-n-decyloxybenzaldehyde, acmc-209g8u, p-decyloxy benzaldehyde
Formula Weight 262.39
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Decyloxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.