Learn More
Selleck Chemical LLC 4-Demethylepipodophyllotoxin(NSC-122819.VM-26) S2254-25mg
SDP
Catalog No. 503108175
Encompass_Preferred
4-Demethylepipodophyllotoxin (NSC-122819 VM-26 DMEP) is an antimitotic agent which binds to monomeric tubulin preventing micro-tubule polymerization It is a potent inhibitor of microtubule assembly
Catalog No. 50-310-8175 Supplier Selleck Chemical LLC Supplier No. S225425MG
4-Demethylepipodophyllotoxin (NSC-122819 VM-26 DMEP) is an antimitotic agent which binds to monomeric tubulin preventing micro-tubule polymerization It is a potent inhibitor of microtubule assembly