Learn More
Medchemexpress LLC 4'-Demethylepipodophyllotoxin | 6559-91-7 | 99.8% | 400.38 | 1 ML
SDP
Catalog No. 5000386194
Encompass_Preferred
4'-Demethylepipodophyllotoxin (4'-DMEP) is an intermediate compound that inhibits microtubule assembly. It is for research use only and not sold to patients. The product appears as a solid, white to off-white substance derived from the plants Berberidaceae Dysosma versipellis (Hance) M. Cheng ex Ying.
  • Inhibits microtubule assembly.
  • Shows cytotoxicity against various tumor cells.
  • Decreases XooFtsZ'GTPase activity and exhibits a bacteriostatic effect.
  • Reduces tumor incidence and tumor volume in mouse skin carcinogenesis models.
  • Stable in powder form for up to 3 years at -20°C.
  • Stable in solvent form for up to 2 years at -80°C.
Catalog No. 50-003-86194 Supplier Medchemexpress LLC Supplier No. HY1743510MM/1ML
Add to Cart
Add to Cart
4'-Demethylepipodophyllotoxin (4'-DMEP) is an intermediate compound that inhibits microtubule assembly. It is for research use only and not sold to patients. The product appears as a solid, white to off-white substance derived from the plants Berberidaceae Dysosma versipellis (Hance) M. Cheng ex Ying.
  • Inhibits microtubule assembly.
  • Shows cytotoxicity against various tumor cells.
  • Decreases XooFtsZ'GTPase activity and exhibits a bacteriostatic effect.
  • Reduces tumor incidence and tumor volume in mouse skin carcinogenesis models.
  • Stable in powder form for up to 3 years at -20°C.
  • Stable in solvent form for up to 2 years at -80°C.