Learn More
TARGETMOL CHEMICALS INC Research Chemical 25mg
SDP
Catalog No. 503038159
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4'-Demethylpodophyllotoxin is isolated from the herb of Dysosma pleiantha (Hance) Woodson with remarkable cytotoxic potential in diverse cancer cell lines. purity: 99%
Catalog No. 50-303-8159 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S002925MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4'-Demethylpodophyllotoxin is isolated from the herb of Dysosma pleiantha (Hance) Woodson with remarkable cytotoxic potential in diverse cancer cell lines. purity: 99%