Learn More
Cayman Chemical 4-DemethylpodophylotoxIn 50mg
SDP
Catalog No. 502911575
Encompass_Preferred
A lignan; inhibits the assembly of isolated chicken brain tubulin (IC50 = 0.5 µM) and the proliferation of LNCaP and PC3 cells (IC50s = 0.056 and 0.082 µM, respectively)
Catalog No. 50-291-1575 Supplier Cayman Chemical Supplier No. 3648150 MG
A lignan; inhibits the assembly of isolated chicken brain tubulin (IC50 = 0.5 µM) and the proliferation of LNCaP and PC3 cells (IC50s = 0.056 and 0.082 µM, respectively)