Learn More
Cayman Chemical 4-deoxy-4α-Phorbol, Purity: ≥98%, CAS: 37415-57-9, 25 mg
SDP
Catalog No. 502933604
Encompass_Preferred
A tetracyclic diterpene found in croton oil; has been used in the semisynthesis of inhibitors of the HIV-1-induced cytopathic effect on MT-4 cells and 4a-phorbol esters
Catalog No. 50-293-3604 Supplier Cayman Chemical Supplier No. 2499825 MG
A tetracyclic diterpene found in croton oil; has been used in the semisynthesis of inhibitors of the HIV-1-induced cytopathic effect on MT-4 cells and 4a-phorbol esters