Learn More
Cayman Chemical 4-deoxy NIvalenol-13C15 1.2mL
SDP
Catalog No. 502991556
Encompass_Preferred
An internal standard for the quantification of 4-deoxy nivalenol by GC- or LC-MS
Catalog No. 50-299-1556 Supplier Cayman Chemical Supplier No. 312491.2 ML
An internal standard for the quantification of 4-deoxy nivalenol by GC- or LC-MS