Learn More
Medchemexpress LLC 4-deoxypyridoxine hydrochloride | 148-51-6 | MFCD00051296 | 99.7% | 189.64 | C8H12ClNO2 | 250 MG
Supplier: Medchemexpress LLC 250MGHYW015210
4-Deoxypyridoxine hydrochloride is the hydrochloride salt of 4-deoxypyridoxine, a vitamin B6 (pyridoxine) antagonist used as a biochemical reagent in life-science research and analytical applications. It is supplied as a high-purity material suitable for biochemical assays, analytical standards, and laboratory experiments.
- Vitamin B6 antagonist for biochemical studies.
- High purity (99.66%) appropriate for analytical and research use.
- Hydrochloride form enhances solubility and handling.
- Available in laboratory-friendly pack sizes such as 250 mg and 1 g.
- Molecular formula C8H12ClNO2; molecular weight 189.64.