Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Deoxypyridoxine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D0053100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0053100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0053100MG Supplier TCI America Supplier No. D0053100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 148-51-6
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.639
MDL Number MFCD00051296
InChI Key QZKKOQQIVLXUEI-UHFFFAOYSA-N
Synonym 5-Hydroxy-4,6-dimethyl-3-pyridinemethanol Hydrochloride
PubChem CID 67417
IUPAC Name 5-(hydroxymethyl)-2,4-dimethylpyridin-3-ol;hydrochloride
SMILES CC1=C(C(=NC=C1CO)C)O.Cl

Specifications

Melting Point 274°C
Color White-Yellow
Quantity 100 mg
Formula Weight 189.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Deoxypyridoxine Hydrochloride
RTECSNumber UT4800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.