Learn More
Cayman Chemical 4-desmethoxy Omeprazole, Purity: ≥98%, CAS: 110374-16-8, 25 mg
SDP
Catalog No. 502987284
Encompass_Preferred
A potential impurity found in commercial omeprazole and esomeprazole magnesium preparations
Catalog No. 50-298-7284 Supplier Cayman Chemical Supplier No. 2212225 MG
A potential impurity found in commercial omeprazole and esomeprazole magnesium preparations