Learn More
Biotium 4-Di-1-ASP
Catalog No. 501964742
Encompass_Preferred
4-Di-1-ASP is a styryl dye stains mitochondria of live cells. It has also been used to stain glioma cells in living brain tissues.Properties: Ex/ Em = 474/606 nm (MeOH); Store at 4°C and protect from light, especially in solution; C16H19IN2; MW: 366.2; [68971-03-9];
Catalog No. 50-196-4742 Supplier Biotium Supplier No. 70006
4-Di-1-ASP is a styryl dye stains mitochondria of live cells. It has also been used to stain glioma cells in living brain tissues.Properties: Ex/ Em = 474/606 nm (MeOH); Store at 4°C and protect from light, especially in solution; C16H19IN2; MW: 366.2; [68971-03-9];