Learn More
Abcam 4-Di-2-ASP, Cationic mitochondrial dye, 5G
Catalog No. 502705350
Encompass_Preferred
MW 394.29 g/mol, Purity >97%. Photostable dye, which is nontoxic to cells and has been employed to stain living nerve terminals in rabbit corneal epithelium, in rat epidermis and at mouse, snake and frog neuromuscular junctions. Methods for using 4-Di-2-ASP to image neuronal cells in live animals have been described.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5350 Supplier Abcam Supplier No. AB1452665G
MW 394.29 g/mol, Purity >97%. Photostable dye, which is nontoxic to cells and has been employed to stain living nerve terminals in rabbit corneal epithelium, in rat epidermis and at mouse, snake and frog neuromuscular junctions. Methods for using 4-Di-2-ASP to image neuronal cells in live animals have been described.
The product is subject to the following: Abcam Restricted Use Statement