Learn More
Medchemexpress LLC 4-Dibenzofurancarboxylic acid | 2786-05-2 | 99.9% | 212.20 | 10 G
SDP
Catalog No. 5000404474
Encompass_Preferred
4-Dibenzofurancarboxylic acid is a synthesis material for polycyclic aromatic hydrocarbons (PAHs) and *Schistosoma mansoni* histone deacetylase 8 (smHDAC8) inhibitors.
  • Solid appearance
  • White to light yellow color
  • Shipped at room temperature
  • Stable for 3 years at room temperature; 2 years in solvent at -80°C, 1 year in solvent at -20°C
Catalog No. 50-004-04474 Supplier Medchemexpress LLC Supplier No. HY3345110G
Add to Cart
Add to Cart
4-Dibenzofurancarboxylic acid is a synthesis material for polycyclic aromatic hydrocarbons (PAHs) and *Schistosoma mansoni* histone deacetylase 8 (smHDAC8) inhibitors.
  • Solid appearance
  • White to light yellow color
  • Shipped at room temperature
  • Stable for 3 years at room temperature; 2 years in solvent at -80°C, 1 year in solvent at -20°C