Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diethylamino)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. D48525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48525G 5 g
D485225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48525G Supplier TCI America Supplier No. D48525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2873-90-7
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.25
MDL Number MFCD00019754
InChI Key KMLGFOAKCYHXCQ-UHFFFAOYSA-N
Synonym 4-diethylamino benzonitrile, p-diethylaminobenzonitrile, benzonitrile, 4-diethylamino, 4-diethylamino benzenecarbonitrile, acmc-1cdnc, maybridge3_004418
PubChem CID 76134
IUPAC Name 4-(diethylamino)benzonitrile
SMILES CCN(CC)C1=CC=C(C=C1)C#N

Specifications

Melting Point 68°C
Quantity 5 g
UN Number 3439
Formula Weight 174.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Diethylamino)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.