Learn More
Medchemexpress LLC HY-W016645 100mg Medchemexpress, 4-Diethylaminobenzaldehyde CAS:120-21-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016645100MG
Encompass_Preferred
Medchemexpress, HY-W016645 100mg 4-Diethylaminobenzaldehyde CAS:120-21-8 4-Diethylaminobenzaldehyde is a reversible aldehyde dehydrogenases (ALDHs) inhibitor, with a Ki of 4 nM for ALDH1. 4-Diethylaminobenzaldehyde displays potent anti-androgenic effect (IC50= 1.71μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3398