Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Difluoromethoxy)benzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D49565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49565G 5 g
D49561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49565G Supplier TCI America Supplier No. D49565G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3447-53-8
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.04
MDL Number MFCD00085007
InChI Key IFNMJBFIRKAQBT-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-difluoromethoxy benzene, 4-difluoromethoxy benzyl bromide, 4-difluoromethoxybenzyl bromide, 4-difluoromethoxy benzylbromide, p-difluoromethoxy benzyl bromide, benzene, 1-bromomethyl-4-difluoromethoxy, 1-bromomethyl-4-difluoromethoxy-benzene, 4-bromomethyl phenoxy difluoromethane, acmc-1aezp, p-difluoromethoxybenzylbromide
PubChem CID 2737001
IUPAC Name 1-(bromomethyl)-4-(difluoromethoxy)benzene
SMILES FC(F)OC1=CC=C(CBr)C=C1

Specifications

Color Yellow
Boiling Point 52°C
Quantity 5 g
UN Number 3265
Formula Weight 237.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Difluoromethoxy)benzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.