Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylamino-1-naphthyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. D16695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16695G 5 g
D16691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16695G Supplier TCI America Supplier No. D16695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29711-79-3
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00003883
InChI Key SMZHGTXTWIAKGS-UHFFFAOYSA-N
Synonym 4-dimethylamino-1-naphthylisothiocyanate, 4-dimethylamino-1-naphthyl isothiocyanate, danito, 4-n,n-dimethylamino-1-naphthylisothiocyanate, n-4-isothiocyanato-1-naphthyl-n,n-dimethylamine, 4-isothiocyanato-n,n-dimethyl-1-naphthalenamine, 4-dimethylamino naphthalenisothiocyanate, acmc-209umu, 4-dimethylamino-1-naphthyl 1sothiocyanate
PubChem CID 122465
IUPAC Name 4-isothiocyanato-N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=C2C=CC=CC2=C(C=C1)N=C=S

Specifications

Melting Point 67°C
Color Yellow
Quantity 5 g
Formula Weight 228.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylamino-1-naphthyl Isothiocyanate [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.