Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)-2-methylazobenzene 96.0+%, TCI America™
SDP

Catalog No. D13511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D13511G Supplier TCI America Supplier No. D13511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54-88-6
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.322
MDL Number MFCD00008306
InChI Key QXTOLYZIJUDOIT-UHFFFAOYSA-N
Synonym N,N-Dimethyl-4-phenylazo-m-toluidine
PubChem CID 5916
IUPAC Name N,N,3-trimethyl-4-phenyldiazenylaniline
SMILES CC1=C(C=CC(=C1)N(C)C)N=NC2=CC=CC=C2

Specifications

Melting Point 68°C
Boiling Point 150°C
Quantity 1 g
Formula Weight 239.32
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)-2-methylazobenzene
RTECSNumber XU5950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.