Learn More
Medchemexpress LLC 4-(dimethylamino)benzaldehyde | 100-10-7 | MFCD00003381 | 99.9% | 149.19 | C9H11NO | 500g
SDP
Catalog No. 5000269381
Encompass_Preferred
p-Dimethylaminobenzaldehyde (4-Dimethylaminobenzaldehyde) can be used as a chromogenic reagent or as an absorbent agent for heavy metal ions[1][2]
Catalog No. 50-002-69381 Supplier Medchemexpress LLC Supplier No. HYY0015500G
p-Dimethylaminobenzaldehyde (4-Dimethylaminobenzaldehyde) can be used as a chromogenic reagent or as an absorbent agent for heavy metal ions[1][2]