Learn More
Medchemexpress LLC 4-(dimethylamino)benzaldehyde | 100-10-7 | MFCD00003381 | 99.9% | 149.19 g/mol | C9H11NO | 250g
SDP
Catalog No. 5000269396
Encompass_Preferred
p-Dimethylaminobenzaldehyde (4-Dimethylaminobenzaldehyde) can be used as a chromogenic reagent or as an absorbent agent for heavy metal ions[1][2]
Catalog No. 50-002-69396 Supplier Medchemexpress LLC Supplier No. HYY0015250G
p-Dimethylaminobenzaldehyde (4-Dimethylaminobenzaldehyde) can be used as a chromogenic reagent or as an absorbent agent for heavy metal ions[1][2]