Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzaldehyde, ≥99% (HPLC), Solstice

Catalog No. 60046742 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-742 50 g Glass Bottle
60-046-741 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-742 Supplier Solstice Supplier No. 3907050G

missing translation for 'validMainframeMsg'

for the determination of hydroxyproline,≥99.0% (HPLC)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 73°C to 75°C
Color White
Boiling Point 176°C to 177°C
Flash Point 164°C (327.2°F)
Ignition Residue ≤0.1% (as SO4)
Loss on Drying 0 to 0.5%
Packaging Glass Bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 50 g
UN Number NONH for all modes of transport
Beilstein 606802
Formula Weight 149.19g/mol
Percent Purity ≥99% (HPLC)
Chemical Name or Material 4-(Dimethylamino)benzaldehyde
Show More Show Less
RTECSNumber 202-819-0
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.