Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzaldehyde, MilliporeSigma™

Catalog No. M1030580100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1030580100 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1030580100 Supplier MilliporeSigma Supplier No. 1.03058.0100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Color Yellow
Density 350kg/m3
Ignition Point 445°C
Packaging Plastic Bottle
Linear Formula 4-[(CH3)CH2N]CCH6HCH4CHO
Quantity 100 g
Solubility Information 0.3g/L (20°C)
Formula Weight 149.19
Grade EP/Reagent
Physical Form Solid
Chemical Name or Material 4-(Dimethylamino)benzaldehyde, For Analysis
Show More Show Less
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.